Assay Details
| Gene Symbol: MIR381HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104192.1 | 2 - 3 | 225 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 655 | MIR655 | 
					MIRN655, hsa-mir-655, mir-655  | 
					-1.4kb | Sense | Chr. 14 - 101049550 - 101049646 on NCBI Build GRCh38 | 
| Downstream | microRNA 487a | MIR487A | 
					MIRN487, MIRN487A, hsa-mir-487, mir-487a  | 
					651.0 | Sense | Chr. 14 - 101052446 - 101052525 on NCBI Build GRCh38 | 
More Information
Set Membership: | 
		   
					
						
							 | 
				

MIR381HG
Genomic Map