Assay Details
| Gene Symbol: LOC101927045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110032.1 | 1 | 118 | 77 | ||
| GenBank mRNA | AK056368.1 | - | 1 | 99 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | short chain dehydrogenase/reductase family 39U member 1 | SDR39U1 |
C14orf124, HCDI |
-48.0 | Anti-sense | Chr. 14 - 24439766 - 24442905 on NCBI Build GRCh38 |
| Downstream | chymase 1 | CMA1 |
CYH, MCT1, chymase |
62.2kb | Anti-sense | Chr. 14 - 24505355 - 24508265 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927045
Genomic Map