Assay Details
| Gene Symbol: LOC101927374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110133.1 | 1 | 221 | 61 | ||
| GenBank mRNA | AK094694.1 | - | 1 | 221 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | roundabout guidance receptor 2 | ROBO2 | SAX3 | -1.8mb | Sense | Chr. 3 - 75906675 - 77649964 on NCBI Build GRCh38 |
| Downstream | microRNA 3923 | MIR3923 | 95.9kb | Sense | Chr. 3 - 79507887 - 79507969 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927374
Genomic Map