Assay Details
Gene Symbol: ATP6V1B1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110273.1 | 3 | 757 | 102 | ||
NR_110274.1 | 2 | 619 | 102 | |||
GenBank mRNA | BM014398.1 | - | 2 | 557 | 102 | |
BM464091.1 | - | 3 | 713 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ventral anterior homeobox 2 | VAX2 | DRES93 | -8.6kb | Anti-sense | Chr. 2 - 70900551 - 70933446 on NCBI Build GRCh38 |
Downstream | ankyrin repeat domain 53 | ANKRD53 | 34.8kb | Anti-sense | Chr. 2 - 70977183 - 70985499 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
