| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000520035.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000520035.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000520035.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000523564.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000523564.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000523564.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000602950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000602950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000602950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012434.hg.1 |
ENST00000602950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000511322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
ENST00000511322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012280.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
TC1700012280.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203039.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
TC1700012280.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203035.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000511322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
ENST00000511322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012280.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203037.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203037.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203037.hg.1 |
TC1700012280.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700203037.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203037.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
TC1700012280.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000502823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000507337.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
ENST00000510360.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
NR_110882.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
NR_110883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700203038.hg.1 |
NR_110884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000520035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000520035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000520035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000523564.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000523564.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000523564.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000602950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000602950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000602950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012434.hg.1 |
ENST00000602950.1 |
Ensembl |