Assay Details
| Gene Symbol: THSD4-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120346.1 | 3 | 1920 | 66 | ||
| GenBank mRNA | AK055527.1 | - | 3 | 1920 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | THSD4 antisense RNA 1 | THSD4-AS1 | -499.6kb | Sense | Chr. 15 - 71166812 - 71189059 on NCBI Build GRCh38 | |
| Downstream | nuclear receptor subfamily 2 group E member 3 | NR2E3 |
ESCS, PNR, RNR, RP37, rd7 |
116.6kb | Anti-sense | Chr. 15 - 71810548 - 71818259 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

THSD4-AS2
Genomic Map