Assay Details
| Gene Symbol: LOC100131626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046369.1 | 4 | 475 | 146 | ||
| NR_046370.1 | 4 | 422 | 146 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitination factor E4A | UBE4A |
E4, UBOX2, UFD2 |
Anti-sense | Chr. 11 - 118359581 - 118399211 on NCBI Build GRCh38 | |
| Downstream |
ATP synthase, H+ transporting, mitochondrial Fo complex subunit G |
ATP5L | ATP5JG | 19.8kb | Anti-sense | Chr. 11 - 118401389 - 118409847 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100131626
Genomic Map