| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007223.hg.1 |
ENST00000433499.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007223.hg.1 |
ENST00000433499.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007223.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007223.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007223.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149553.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149553.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149553.hg.1 |
TC0X00007223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007223.hg.1 |
ENST00000433499.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007223.hg.1 |
ENST00000433499.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007223.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007223.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007223.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149555.hg.1 |
ENST00000433499.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149555.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149555.hg.1 |
ENST00000433499.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149555.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149555.hg.1 |
TC0X00007223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00149554.hg.1 |
ENST00000433499.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149554.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149554.hg.1 |
ENST00000433499.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149554.hg.1 |
NR_046649.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149554.hg.1 |
TC0X00007223.hg.1 |
Clariom D TC |