Assay Details
| Gene Symbol: ERI3-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046817.1 | 2 | 625 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ERI1 exoribonuclease family member 3 | ERI3 |
PINT1, PRNPIP |
Sense | Chr. 1 - 44221070 - 44355298 on NCBI Build GRCh38 | |
| Downstream | ring finger protein 220 | RNF220 | C1orf164 | 160.8kb | Anti-sense | Chr. 1 - 44404846 - 44651724 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

ERI3-IT1
Genomic Map