| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
ENST00000509367.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
ENST00000509367.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
ENST00000627386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008416.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157629.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157629.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157629.hg.1 |
TC0500008416.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500157614.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157614.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157614.hg.1 |
TC0500008416.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
ENST00000509367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
ENST00000509367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
ENST00000627386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008416.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157618.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157618.hg.1 |
ENST00000627386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157618.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157618.hg.1 |
ENST00000509669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157618.hg.1 |
ENST00000627386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157618.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157618.hg.1 |
TC0500008416.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500157628.hg.1 |
ENST00000509367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157628.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157628.hg.1 |
ENST00000509367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157628.hg.1 |
NR_104997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157628.hg.1 |
TC0500008416.hg.1 |
Clariom D TC |