| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012071.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012071.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012071.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012071.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012071.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012071.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183568.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183568.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183568.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183568.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183568.hg.1 |
TC0500012071.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500012071.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012071.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012071.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012071.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012071.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012071.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183570.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183570.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183570.hg.1 |
ENST00000512748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183570.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183570.hg.1 |
TC0500012071.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500183572.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183572.hg.1 |
NR_105044.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183572.hg.1 |
TC0500012071.hg.1 |
Clariom D TC |