Assay Details
| Gene Symbol: LOC101928069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120586.1 | 4 | 748 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein L11 | MRPL11 |
CGI-113, L11MT, MRP-L11 |
-34.6kb | Sense | Chr. 11 - 66435075 - 66438860 on NCBI Build GRCh38 |
| Downstream | dipeptidyl peptidase 3 | DPP3 | DPPIII | 6.1kb | Anti-sense | Chr. 11 - 66480013 - 66509659 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186892.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186892.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186892.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186892.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186892.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186892.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186892.hg.1 | TC1100011289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186903.hg.1 | TC1100011289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | ENST00000525142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | ENST00000527274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186904.hg.1 | TC1100011289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100186891.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186891.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186891.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186891.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186891.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186891.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186891.hg.1 | TC1100011289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100186898.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186898.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186898.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186898.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186898.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186898.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186898.hg.1 | ENST00000533502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186898.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186898.hg.1 | TC1100011289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100186890.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186890.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186890.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186890.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186890.hg.1 | ENST00000527092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100186890.hg.1 | NR_120586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100186890.hg.1 | TC1100011289.hg.1 | Clariom D TC |

LOC101928069
Genomic Map