Assay Details
| Gene Symbol: RXFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253730.1 | NP_001240659.1 | 2 | 452 | 62 | |
| GenBank mRNA | BC143559.1 | - | 2 | 274 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001253730.1 transcripts.
Set Membership: |
|

RXFP1
Genomic Map