Assay Details
| Gene Symbol: EMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001423.2 | NP_001414.1 | 5 | 2423 | 214 | |
| GenBank mRNA | BC047300.1 | - | 5 | 2348 | 4444217 | 214 |
| U77085.1 | - | 5 | 2392 | 214 | ||
| Y07909.1 | - | 5 | 2394 | 214 | ||
| Z50751.1 | - | 5 | 2298 | 214 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000189.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000535134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000535134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002396.hg.1 | ENST00000256951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002396.hg.1 | ENST00000537612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002396.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002396.hg.1 | TC12000189.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200148346.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200148346.hg.1 | ENST00000537612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200148346.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200148346.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200148346.hg.1 | ENST00000537612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200148346.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200148346.hg.1 | TC1200006909.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000535134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000535134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 198606 | - | - |

EMP1
Genomic Map