Assay Details
| Gene Symbol: CDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001804.2 | NP_001795.2 | 2 - 3 | 685 | 120 | |
| GenBank mRNA | AF239666.1 | - | 2 - 3 | 670 | 120 | |
| BC096251.1 | - | 3 - 4 | 302 | 120 | ||
| BC096252.1 | - | 3 - 4 | 348 | 40005955 | 120 | |
| BC127122.1 | - | 3 - 4 | 348 | 40005957 | 120 | |
| U15212.1 | - | 2 - 3 | 403 | 120 | ||
| U51095.1 | - | 2 - 3 | 670 | 120 | ||
More Information
Set Membership: |
|

CDX1
Genomic Map