Assay Details
| Gene Symbol: AFAP1-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AFAP1-NTRK2_A14N12.0 | - | 77 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AFAP1-NTRK2 fusion| Gene Symbol: AFAP1-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AFAP1-NTRK2_A14N12.0 | - | 77 | 82 | ||