Assay Details
| Gene Symbol: ATIC-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ATIC-ALK_A7A20.0 | - | 122 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATIC-ALK fusion| Gene Symbol: ATIC-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ATIC-ALK_A7A20.0 | - | 122 | 82 | ||