Assay Details
| Gene Symbol: AXL-MBIP fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AXL-MBIP_A20M4_1.0 | - | 89 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AXL-MBIP fusion| Gene Symbol: AXL-MBIP fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AXL-MBIP_A20M4_1.0 | - | 89 | 92 | ||