Assay Details
| Gene Symbol: BAG4-FGFR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BAG4-FGFR1_B1F8.0 | - | 55 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BAG4-FGFR1 fusion| Gene Symbol: BAG4-FGFR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BAG4-FGFR1_B1F8.0 | - | 55 | 105 | ||
Set Membership: |
|