Assay Details
| Gene Symbol: KTN1-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KTN1-RET_K29R12_COSF1513.0 | - | 49 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KTN1-RET fusion| Gene Symbol: KTN1-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KTN1-RET_K29R12_COSF1513.0 | - | 49 | 93 | ||