Assay Details
| Gene Symbol: OXR1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_OXR1-MET_O9M13.0 | - | 46 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OXR1-MET fusion| Gene Symbol: OXR1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_OXR1-MET_O9M13.0 | - | 46 | 89 | ||