Assay Details
| Gene Symbol: QKI-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_QKI-NTRK2_Q6N16.0 | - | 64 | 117 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

QKI-NTRK2 fusion| Gene Symbol: QKI-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_QKI-NTRK2_Q6N16.0 | - | 64 | 117 | ||
Set Membership: |
|