Assay Details
| Gene Symbol: PI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002638.3 | NP_002629.1 | 2 - 3 | 385 | 84 | |
| GenBank mRNA | AA583773.1 | - | 1 - 2 | 84 | ||
| BC010952.1 | - | 2 - 3 | 386 | 4083155 | 84 | |
| Z18538.1 | - | 2 - 3 | 363 | 84 | ||
More Information
Set Membership: |
|

PI3
Genomic Map