Assay Details
| Gene Symbol: HEY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282851.1 | NP_001269780.1 | 1 - 2 | 270 | 86 | |
| GenBank mRNA | AK293646.1 | - | 1 - 2 | 270 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293646.1, NM_001282851.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
pulmonary valve morphogenesis
atrioventricular valve formation
epithelial to mesenchymal transition involved in endocardial cushion formation
endocardial cushion morphogenesis
cardiac ventricle morphogenesis
transcription, DNA-templated
Notch signaling pathway
dorsal aorta morphogenesis
umbilical cord morphogenesis
negative regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cardiac epithelial to mesenchymal transition
heart trabecula formation
cardiac septum morphogenesis
ventricular septum morphogenesis
labyrinthine layer blood vessel development
arterial endothelial cell differentiation
Notch signaling involved in heart development
cellular response to glucocorticoid stimulus
negative regulation of transcription regulatory region DNA binding
negative regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
regulation of vasculogenesis
transcription factor activity, RNA polymerase II core promoter sequence-specific
transcription factor activity, protein binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
microsatellite binding
protein dimerization activity
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
pulmonary valve morphogenesis
atrioventricular valve formation
epithelial to mesenchymal transition involved in endocardial cushion formation
endocardial cushion morphogenesis
cardiac ventricle morphogenesis
transcription, DNA-templated
Notch signaling pathway
dorsal aorta morphogenesis
umbilical cord morphogenesis
negative regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cardiac epithelial to mesenchymal transition
heart trabecula formation
cardiac septum morphogenesis
ventricular septum morphogenesis
labyrinthine layer blood vessel development
arterial endothelial cell differentiation
Notch signaling involved in heart development
cellular response to glucocorticoid stimulus
negative regulation of transcription regulatory region DNA binding
negative regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
regulation of vasculogenesis
transcription factor activity, RNA polymerase II core promoter sequence-specific
transcription factor activity, protein binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
microsatellite binding
protein dimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000337919.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000354724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000435063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000435063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000435063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000435063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000435063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000518733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000518733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000518733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000518733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000518733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000519075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000519075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000519075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000519075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000519075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000519075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000519075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000521111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000521111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000521111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000521111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000521111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000521111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523976.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523976.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523976.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523976.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523976.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | ENST00000523976.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001040708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001282851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001282851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001282851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001282851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001282851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001282851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001282851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_001282851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010894.hg.1 | NM_012258.3 | Refseq |

HEY1
Genomic Map