Assay Details
| Gene Symbol: LRRFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005265551.1 | XP_005265608.1 | 7 - 8 | 719 | 103 | |
| XM_006713400.1 | XP_006713463.1 | 5 - 6 | 611 | 103 | ||
| XM_006713404.1 | XP_006713467.1 | 5 - 6 | 611 | 103 | ||
| XM_011534229.1 | XP_011532531.1 | 6 - 7 | 654 | 103 | ||
| XM_017007471.1 | XP_016862960.1 | 6 - 7 | 668 | 103 | ||
| XM_017007472.1 | XP_016862961.1 | 7 - 8 | 719 | 103 | ||
| XM_017007473.1 | XP_016862962.1 | 6 - 7 | 668 | 103 | ||
| XM_017007474.1 | XP_016862963.1 | 6 - 7 | 668 | 103 | ||
| XM_017007475.1 | XP_016862964.1 | 6 - 7 | 1400 | 103 | ||
| XM_017007477.1 | XP_016862966.1 | 6 - 7 | 1400 | 103 | ||
| XM_017007479.1 | XP_016862968.1 | 6 - 7 | 668 | 103 | ||
More Information
Set Membership: |
|

LRRFIP2
Genomic Map