Assay Details
| Gene Symbol: ZDHHC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK310561.1 | - | 3 - 4 | 970 | 64 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000525925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000525925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000525925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | ENST00000602427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | NM_207340.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | NM_207340.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | NM_207340.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | NM_207340.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011291.hg.1 | NM_207340.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000310442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000525925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000525925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000525925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000526986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000534073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | ENST00000602427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | NM_207340.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | NM_207340.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | NM_207340.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | NM_207340.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011291.hg.1 | NM_207340.2 | Refseq |

ZDHHC24
Genomic Map