Assay Details
| Gene Symbol: BAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172415.1 | NP_001165886.1 | 4 - 5 | 868 | 105 | |
| NM_004323.5 | NP_004314.5 | 4 - 5 | 868 | 105 | ||
| GenBank mRNA | AF022224.1 | - | 4 - 5 | 826 | 105 | |
| AF116273.1 | - | 4 - 5 | 796 | 105 | ||
| AK093238.1 | - | 1 - 2 | 1527 | 105 | ||
| AK096038.1 | - | 5 - 6 | 631 | 105 | ||
| AK222749.1 | - | 4 - 5 | 815 | 105 | ||
| AK316577.1 | - | 4 - 5 | 794 | 105 | ||
| BC001936.1 | - | 4 - 5 | 829 | 2823774 | 105 | |
| BC014774.1 | - | 4 - 5 | 678 | 105 | ||
| KU177932.1 | - | 4 - 5 | 568 | 105 | ||
| KU177933.1 | - | 5 - 6 | 710 | 105 | ||
| U46917.1 | - | 4 - 5 | 828 | 105 | ||
| Z35491.1 | - | 4 - 5 | 846 | 105 | ||
More Information
Set Membership: |
|

BAG1
Genomic Map