| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |