Assay Details
| Gene Symbol: LINC01589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131244.1 | 1 - 2 | 246 | 70 | ||
| GenBank mRNA | BC045163.1 | - | 1 - 2 | 220 | 70 | |
| BQ188486.1 | - | 1 - 2 | 246 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibulin 1 | FBLN1 |
FBLN, FIBL1 |
-3.3kb | Anti-sense | Chr. 22 - 45502839 - 45601134 on NCBI Build GRCh38 |
| Downstream | ataxin 10 | ATXN10 |
E46L, HUMEEP, SCA10 |
66.2kb | Anti-sense | Chr. 22 - 45671798 - 45845307 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01589
Genomic Map