Assay Details
| Gene Symbol: LINC01601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131768.1 | 3 - 4 | 390 | 167 | ||
| GenBank mRNA | BX119171.1 | - | 3 - 4 | 390 | 167 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1478 | LINC01478 | -11.2kb | Anti-sense | Chr. 18 - 44323435 - 44531697 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101927943 | LOC101927943 | 123.8kb | Anti-sense | Chr. 18 - 44676927 - 44679717 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01601
Genomic Map