Assay Details
| Gene Symbol: PRRX2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132102.1 | 1 - 2 | 134 | 92 | ||
| GenBank mRNA | BX110190.1 | - | 1 - 2 | 106 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat and SOCS box containing 6 | ASB6 | -70.7kb | Sense | Chr. 9 - 129634604 - 129642169 on NCBI Build GRCh38 | |
| Downstream | prostaglandin E synthase | PTGES |
MGST-IV, MGST1-L1, MGST1L1, MPGES, PGES, PIG12, PP102, PP1294, TP53I12, mPGES-1 |
19.6kb | Sense | Chr. 9 - 129738336 - 129777579 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

PRRX2-AS1
Genomic Map