Assay Details
| Gene Symbol: DPY19L1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132360.1 | 5 - 6 | 833 | 85 | ||
| GenBank mRNA | DQ287935.1 | - | 5 - 6 | 834 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 997 | LINC00997 | -71.8kb | Sense | Chr. 7 - 32758286 - 32762924 on NCBI Build GRCh38 | |
| Downstream | kelch repeat and BTB domain containing 2 | KBTBD2 | BKLHD1 | 29.4kb | Anti-sense | Chr. 7 - 32868166 - 32892144 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008207.hg.1 | ENST00000560803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008207.hg.1 | ENST00000560803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000310974.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000428054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000428054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000446375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000462134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000463538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000463538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000463538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000481923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000493989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000493989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000493989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000612226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000612226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000612226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | ENST00000612226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010716.hg.1 | NM_015283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008995.hg.1 | ENST00000411856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000550380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008243.hg.1 | ENST00000553247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008207.hg.1 | ENST00000560803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008207.hg.1 | ENST00000560803.1 | Ensembl |

DPY19L1P2
Genomic Map