Assay Details
| Gene Symbol: GRPEL2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132366.1 | 1 - 2 | 117 | 102 | ||
| GenBank mRNA | AI857303.1 | - | 1 - 2 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
GrpE like 2, mitochondrial |
GRPEL2 | Mt-GrpE#2 | Anti-sense | Chr. 5 - 149345414 - 149354583 on NCBI Build GRCh38 | |
| Downstream | prenylcysteine oxidase 1 like | PCYOX1L | 365.0 | Anti-sense | Chr. 5 - 149358007 - 149375112 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

GRPEL2-AS1
Genomic Map