Assay Details
| Gene Symbol: LOC105379192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134246.1 | 1 - 2 | 68 | 83 | ||
| GenBank mRNA | BX096849.1 | - | 1 - 2 | 68 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transient receptor potential cation channel subfamily C member 7 | TRPC7 | TRP7 | -762.7kb | Anti-sense | Chr. 5 - 136213311 - 136365476 on NCBI Build GRCh38 |
| Downstream | kelch like family member 3 | KLHL3 | PHA2D | 486.5kb | Anti-sense | Chr. 5 - 137617500 - 137736090 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105379192
Genomic Map