Assay Details
| Gene Symbol: LOC105379030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134252.1 | 1 - 2 | 179 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 171 | TMEM171 | PRP2 | -191.0 | Anti-sense | Chr. 5 - 73120561 - 73131817 on NCBI Build GRCh38 |
| Downstream | transmembrane protein 174 | TMEM174 | 22.6kb | Anti-sense | Chr. 5 - 73173196 - 73175143 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105379030
Genomic Map