Assay Details
| Gene Symbol: LOC101927278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134451.1 | 1 - 2 | 437 | 75 | ||
| GenBank mRNA | CB123723.1 | - | 1 - 2 | 112 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4685 | MIR4685 | -15.0kb | Anti-sense | Chr. 10 - 98431292 - 98431360 on NCBI Build GRCh38 | |
| Downstream | heparanase 2 (inactive) | HPSE2 |
HPA2, HPR2, UFS, UFS1 |
7.7kb | Anti-sense | Chr. 10 - 98457077 - 99235875 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927278
Genomic Map