Assay Details
| Gene Symbol: LOC105376468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134496.1 | 2 - 3 | 175 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | membrane palmitoylated protein 7 | MPP7 | -129.9kb | Sense | Chr. 10 - 28050993 - 28303066 on NCBI Build GRCh38 | |
| Downstream | WAC antisense RNA 1 (head to head) | WAC-AS1 | 83.7kb | Sense | Chr. 10 - 28519917 - 28532354 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105376468
Genomic Map