Assay Details
| Gene Symbol: LOC105373006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134617.1 | 3 - 4 | 461 | 63 | ||
| GenBank mRNA | DB089380.1 | - | 3 - 4 | 461 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synapsin III | SYN3 | -52.0kb | Anti-sense | Chr. 22 - 32508022 - 33058391 on NCBI Build GRCh38 | |
| Downstream | LARGE xylosyl- and glucuronyltransferase 1 | LARGE1 |
LARGE, MDC1D, MDDGA6, MDDGB6 |
45.9kb | Anti-sense | Chr. 22 - 33162237 - 33922841 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105373006
Genomic Map