Assay Details
| Gene Symbol: LOC105369785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135022.1 | 1 - 2 | 222 | 73 | ||
| GenBank mRNA | BX484943.1 | - | 1 - 2 | 223 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATP23 metallopeptidase and ATP synthase assembly factor homolog | ATP23 |
KUB3, XRCC6BP1 |
-130.6kb | Sense | Chr. 12 - 57941504 - 57957269 on NCBI Build GRCh38 |
| Downstream | endogenous retrovirus group K member 8 Pol protein-like | LOC105369972 | 235.2kb | Anti-sense | Chr. 12 - 58323535 - 58331743 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105369785
Genomic Map