Assay Details
| Gene Symbol: LOC105370177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135319.1 | 1 - 2 | 84 | 106 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | A-kinase anchoring protein 11 | AKAP11 |
AKAP-11, AKAP220, PPP1R44, PRKA11 |
-101.6kb | Sense | Chr. 13 - 42271470 - 42323267 on NCBI Build GRCh38 |
| Downstream | tumor necrosis factor superfamily member 11 | TNFSF11 |
CD254, ODF, OPGL, OPTB2, RANKL, TNLG6B, TRANCE, hRANKL2, sOdf |
127.4kb | Sense | Chr. 13 - 42562736 - 42608013 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105370177
Genomic Map