Assay Details
| Gene Symbol: LOC105377143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135533.1 | 1 - 2 | 111 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeats and immunoglobulin like domains 1 | LRIG1 |
LIG-1, LIG1 |
-231.7kb | Sense | Chr. 3 - 66378797 - 66501124 on NCBI Build GRCh38 |
| Downstream | kelch repeat and BTB domain containing 8 | KBTBD8 |
TA-KRP, TAKRP |
260.0kb | Anti-sense | Chr. 3 - 66998303 - 67011210 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105377143
Genomic Map