Assay Details
| Gene Symbol: LOC101928583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135555.1 | 2 - 3 | 218 | 81 | ||
| GenBank mRNA | HY157714.1 | - | 2 - 3 | 203 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 7 member 14 | SLC7A14 | PPP1R142 | Anti-sense | Chr. 3 - 170459554 - 170586074 on NCBI Build GRCh38 | |
| Downstream | ribosomal protein L22 like 1 | RPL22L1 | 202.8kb | Anti-sense | Chr. 3 - 170864875 - 170870256 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928583
Genomic Map