| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010812.hg.1 |
ENST00000526931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010812.hg.1 |
ENST00000526931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010812.hg.1 |
ENST00000526931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000436677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000436677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000436677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000443068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011567.hg.1 |
ENST00000443068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013568.hg.1 |
ENST00000502460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013568.hg.1 |
ENST00000502460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013568.hg.1 |
ENST00000502460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007854.hg.1 |
ENST00000391266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007854.hg.1 |
ENST00000419595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007854.hg.1 |
ENST00000419595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007854.hg.1 |
ENST00000419595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007854.hg.1 |
ENST00000419595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007854.hg.1 |
ENST00000423746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007854.hg.1 |
ENST00000423746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007854.hg.1 |
ENST00000423746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007854.hg.1 |
ENST00000423746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013568.hg.1 |
ENST00000502460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013568.hg.1 |
ENST00000502460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013568.hg.1 |
ENST00000502460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007816.hg.1 |
ENST00000414608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000434037.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000436677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000436677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000436677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000443068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011567.hg.1 |
ENST00000443068.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007854.hg.1 |
ENST00000391266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007854.hg.1 |
ENST00000419595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007854.hg.1 |
ENST00000419595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007854.hg.1 |
ENST00000419595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007854.hg.1 |
ENST00000419595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007854.hg.1 |
ENST00000423746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007854.hg.1 |
ENST00000423746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007854.hg.1 |
ENST00000423746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007854.hg.1 |
ENST00000423746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010812.hg.1 |
ENST00000526931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010812.hg.1 |
ENST00000526931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010812.hg.1 |
ENST00000526931.2 |
Ensembl |