Assay Details
| Gene Symbol: LOC102723886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135822.1 | 3 - 4 | 432 | 104 | ||
| GenBank mRNA | AW292250.1 | - | 1 - 2 | 104 | ||
| BE466517.1 | - | 1 - 2 | 104 | |||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BF216856.1 | - | 191 | Chr. - - [-] on Build | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929713 | LOC101929713 | Anti-sense | Chr. 1 - 219218524 - 219445121 on NCBI Build GRCh38 | ||
| Downstream | solute carrier family 30 member 10 | SLC30A10 |
HMDPC, HMNDYT1, ZNT10, ZNT8, ZRC1, ZnT-10 |
473.2kb | Sense | Chr. 1 - 219910398 - 219959754 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC102723886
Genomic Map