Assay Details
| Gene Symbol: LOC105374620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136207.1 | 3 - 4 | 355 | 88 | ||
| GenBank mRNA | HY017505.1 | - | 3 - 4 | 355 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 5 open reading frame 38 | C5orf38 |
CEI, IRX2NB |
-160.4kb | Sense | Chr. 5 - 2751940 - 2758576 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1377 | LINC01377 | 257.3kb | Sense | Chr. 5 - 3177835 - 3181232 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105374620
Genomic Map