| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008875.hg.1 |
ENST00000563087.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008875.hg.1 |
ENST00000563087.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008875.hg.1 |
ENST00000563087.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008875.hg.1 |
ENST00000563087.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008875.hg.1 |
ENST00000564394.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008875.hg.1 |
ENST00000564394.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008875.hg.1 |
ENST00000563087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008875.hg.1 |
ENST00000563087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008875.hg.1 |
ENST00000563087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008875.hg.1 |
ENST00000563087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008875.hg.1 |
ENST00000564394.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008875.hg.1 |
ENST00000564394.1 |
Ensembl |