Assay Details
| Gene Symbol: MIR100HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_137178.1 | 4 - 5 | 1015 | 62 | ||
| GenBank mRNA | BC089451.1 | - | 4 - 5 | 623 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sortilin related receptor 1 | SORL1 |
C11orf32, LR11, LRP9, SORLA, SorLA-1, gp250 |
-394.6kb | Anti-sense | Chr. 11 - 121452203 - 121633762 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000524376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000526674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000526674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000527474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000528986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000529823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000530072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000530072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000530072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000530072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000530072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000530072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000530955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000530955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000531629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000532890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000533109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000533109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000533109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000533109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000534782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000560104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000560104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000560104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000560104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000560104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | ENST00000560104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NM_001001786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NM_001001786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NM_001001786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NM_001001786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NM_001001786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NM_001001786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012583.hg.1 | NR_024430.2 | Refseq |

MIR100HG
Genomic Map