Assay Details
| Gene Symbol: MIR100HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_137190.1 | 1 - 2 | 786 | 80 | ||
| GenBank mRNA | DA532588.1 | - | 1 - 2 | 240 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 125b-1 | MIR125B1 |
MIRN125B1, mir-125b-1 |
-530.0 | Sense | Chr. 11 - 122099757 - 122099844 on NCBI Build GRCh38 |
| Downstream |
BH3-like motif containing, cell death inducer |
BLID | BRCC2 | 13.1kb | Sense | Chr. 11 - 122115354 - 122116215 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MIR100HG
Genomic Map