Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tudor domain containing 5 | TDRD5 | TUDOR3 | -50.7kb | Anti-sense | Chr. 1 - 179591613 - 179691272 on NCBI Build GRCh38 |
| Downstream | torsin 1A interacting protein 2 | TOR1AIP2 |
IFRG15, LULL1, NET9 |
97.3kb | Sense | Chr. 1 - 179839967 - 179878614 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map