| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000589218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011721.hg.1 |
ENST00000591485.5 |
Ensembl |